2-Mercaptobenzimidazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Benzimidazole-2-thiol, 1,3-Dihydro-2H-benzo[d]imidazole-2-thione, Rabeprazole Related Compound C, 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol
M-Clarity™ quality level = MQ200
M3205-500G
M3205-100G
M3205-5G
SIALM3205-500GEA
2690
CZK
SIALM3205-500G
SIALM3205-100G
SIALM3205-5G
2-Mercaptobenzimidazole, Sigma-Aldrich®
2-Mercaptobenzimidazole
Vzorec:
C₇H₆N₂S MW: 150,2 g/mol Bod varu: 376 °C (1013 hPa) Bod tání: 154 °C Hustota: 1,42 g/cm³ (20 °C) |
MDL Number:
MFCD00466107 CAS číslo: 583-39-1 EINECS: 209-502-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...