2-Iodo-1-methylimidazole ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Iodo-1-methyl-1H-imidazole
H51862.MD
H51862.03
H51862.MDEA
1830
CZK
H51862.MD
H51862.03
2-Iodo-1-methylimidazole ≥97%
2-Iodo-1-methylimidazole
Purity: 97%
CAS No.: 37067-95-1
Molecular Formula: C4H5IN2
Formula Weight: 208.00
Melting Point: 79-82°
MDL No.: MFCD00661606
Vzorec:
C₄H₅IN₂ Bod tání: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00661606 CAS číslo: 37067-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...