Tisk…

2-Hydroxybenzimidazole ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-Benzimidazolinone, 2(1H)-Benzimidazolone, 2-Hydroxy-1H-benzimidazole

A18324.09 A18324.18 A18324.22 A18324.30
A18324.09EA 1290 CZK
A18324.09 A18324.18 A18324.22 A18324.30
2-Hydroxybenzimidazole ≥98%
2-Hydroxybenzimidazole

Purity: 98% CAS No.: 615-16-7 Molecular Formula: C7H6N2O Formula Weight: 134.14 Melting Point: 314-316° Beilstein Reference: 119866

Vzorec: C₇H₆N₂O
MW: 134.14
Bod tání: 314…316 °C
Hustota: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00127894
CAS číslo: 615-16-7
EINECS: 210-412-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR