2-Fluoro-5-nitrobenzaldehyde ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
L19233.03
L19233.06
L19233.03EA
1220
CZK
L19233.03
L19233.06
2-Fluoro-5-nitrobenzaldehyde ≥98%
2-Fluoro-5-nitrobenzaldehyde
Purity: 98%
CAS No.: 27996-87-8
Molecular Formula: C7H4FNO3
Formula Weight: 169.11
Melting Point: 57-59°
Beilstein Reference: 1950329
Vzorec:
C₇H₄FNO₃ MW: 169,11 g/mol Bod tání: 57…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042298 CAS číslo: 27996-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...