Tisk…

2-Fluoro-4-methylbenzaldehyde ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Fluoro-p-tolylaldehyde, 2-Fluoro-4-tolualdehyde, 2-Fluoro-p-tolualdehyde

H26626.03 H26626.06
H26626.03EA 2900 CZK
H26626.03 H26626.06
2-Fluoro-4-methylbenzaldehyde ≥97%
2-Fluoro-4-methylbenzaldehyde

Purity: 97% CAS No.: 146137-80-6 Molecular Formula: C8H7FO Formula Weight: 138.14 Refractive Index: 1.5240

Vzorec: C₈H₇FO
MW: 138,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094325
CAS číslo: 146137-80-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR