Tisk…

Ethyl-2-ethylacetoacetate 95%

Dodavatel: Thermo Fisher Scientific
Varování

B22295.14 B22295.22
B22295.14EA 3260 CZK
B22295.14 B22295.22
Ethyl-2-ethylacetoacetate 95%
Ethyl-2-ethylacetoacetate

Purity: 95% CAS No.: 607-97-6 Molecular Formula: C8H14O3 Formula Weight: 158.20 Flash Point: 77°(170°F) Refractive Index: 1.4220 Boiling Point: 83-85°/10mm Beilstein Reference: 636286 MDL No.: MFCD00039898

Vzorec: C₈H₁₄O₃
Bod varu: 83…85 °C (10 mmHg)
Hustota: 1,009
Bod vzplanutí: 77 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039898
CAS číslo: 607-97-6
EINECS: 210-151-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR