2-Acetylfuran, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Furyl methyl ketone, 1-(Furan-2-yl)ethanone
M-Clarity™ quality level = MQ100
A16254-100G
A16254-25G
SIALA16254-100GEA
1710
CZK
SIALA16254-100G
SIALA16254-25G
2-Acetylfuran, Sigma-Aldrich®
2-Acetylfuran
Vzorec:
C₆H₆O₂ MW: 110.11 Bod varu: 183 Bod tání: 29…33 °C Hustota: 1.098 g/cm³ (20 °C) |
MDL Number:
MFCD00003242 CAS číslo: 1192-62-7 EINECS: 214-757-1 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...