2-Acetyl-1-naphthol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1'-Hydroxy-2'-acetonaphthone, 1-(1-Hydroxynaphthalen-2-yl)ethan-1-one, 2-Acetyl-1-hydroxynaphthalene, 1-Hydroxy-2-acetonaphthone
A19532.14
A19532.22
A19532.14EA
2070
CZK
A19532.14
A19532.22
2-Acetyl-1-naphthol ≥99%
2-Acetyl-1-naphthol
Purity: 99%
CAS No.: 711-79-5
Molecular Formula: C12H10O2
Formula Weight: 186.21
Melting Point: 98-100°
Beilstein Reference: 1868084
Vzorec:
C₁₂H₁₀O₂ Bod varu: 110…115 °C (0,5 mmHg) Bod tání: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003963 CAS číslo: 711-79-5 EINECS: 211-918-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...