Tisk…

2-Furonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Cyanofuran

M-Clarity™ quality level = MQ100

159573-25G 159573-5G
SIAL159573-25GEA 14330 CZK
SIAL159573-25G SIAL159573-5G
2-Furonitrile, Sigma-Aldrich®
2-Furonitrile
Vzorec: C₅H₃NO
MW: 93,08 g/mol
Bod varu: 146 °C (1013 hPa)
Hustota: 1,085 g/cm³ (20 °C)
MDL Number: MFCD00003223
CAS číslo: 617-90-3
EINECS: 210-537-4
UN: 1992
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR