Tisk…

2-Cyano-4-methylpyridine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Methylpyridine-2-carbonitrile, 4-Methylpicolinonitrile, 2-Cyano-γ-picoline, 2-Cyano-4-picoline, 4-Methyl-2-pyridinecarbonitrile

M-Clarity™ quality level = MQ100

673463-1G 673463-5G
SIAL673463-1GEA 3690 CZK
SIAL673463-1G SIAL673463-5G
2-Cyano-4-methylpyridine, Sigma-Aldrich®
2-Cyano-4-methylpyridine
Vzorec: C₇H₆N₂
MW: 118,14 g/mol
Bod varu: 145…148 °C (38 mmHg)
Bod tání: 90…91 °C
MDL Number: MFCD00128868
CAS číslo: 1620-76-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR