Tisk…

2-Cyano-4-methylpyridine ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Methylpyridine-2-carbonitrile, 4-Methylpicolinonitrile, 2-Cyano-γ-picoline, 2-Cyano-4-picoline, 4-Methyl-2-pyridinecarbonitrile

L17148.03 L17148.06
L17148.03EA 2380 CZK
L17148.03 L17148.06
2-Cyano-4-methylpyridine ≥98%
2-Cyano-4-methylpyridine

Purity: 98% CAS No.: 1620-76-4 Molecular Formula: C7H6N2 Formula Weight: 118.14 Odour: Characteristic Melting Point: 85-88° Boiling Point: 145-148°/38mm Beilstein Reference: 110753 MDL No.: MFCD00128868

Vzorec: C₇H₆N₂
MW: 118,14 g/mol
Bod varu: 145…148 °C (38 mmHg)
Bod tání: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00128868
CAS číslo: 1620-76-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR