2-Cyano-3-methylpyridine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L19456.06
L19456.14
L19456.22
L19456.06EA
1210
CZK
L19456.06
L19456.14
L19456.22
2-Cyano-3-methylpyridine ≥98%
2-Cyano-3-methylpyridine
Purity: 98%
CAS No.: 20970-75-6
Molecular Formula: C7H6N2
Formula Weight: 118.14
Flash Point: 115°(239°F)
Odour: Characteristic
Boiling Point: 141°/38mm
Beilstein Reference: 112001
MDL No.: MFCD00190585
Vzorec:
C₇H₆N₂ Bod varu: 141 °C (38 mmHg) Bod tání: 84…87 °C Bod vzplanutí: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190585 CAS číslo: 20970-75-6 EINECS: 244-131-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...