Tisk…

2-Chlorothanesulphonyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (2-Chloroethyl)sulphonyl chloride

A15377.30 A15377.09 A15377.18 A15377.22
A15377.30EA 22620 CZK
A15377.30 A15377.09 A15377.18 A15377.22
2-Chlorothanesulphonyl chloride ≥98%
2-Chloroethanesulphonyl chloride

Purity: 98% CAS No.: 1622-32-8 Molecular Formula: C2H4Cl2O2S Formula Weight: 163.02 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.4915 Boiling Point: 203° Beilstein Reference: 1751202 UN No.: 3390 MDL No.: MFCD00007461

Vzorec: C₂H₄Cl₂O₂S
MW: 163.02
Bod varu: 203 °C
Bod tání: 3
Hustota: 1.560
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007461
CAS číslo: 1622-32-8
EINECS: 216-594-1
UN: 3390
ADR: 6.1 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR