Tisk…

2-Chlorobenzimidazole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

592277-25G 592277-5G
SIAL592277-25GEA 9150 CZK
SIAL592277-25G SIAL592277-5G
2-Chlorobenzimidazole, Sigma-Aldrich®
2-Chlorobenzimidazole
Vzorec: C₇H₅ClN₂
MDL Number: MFCD00051944
CAS číslo: 4857-06-1
EINECS: 225-453-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR