2-Chloro-4-cyanobenzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Chloro-4-cyanobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
638870-1G
638870-5G
SIAL638870-1GEA
2470
CZK
SIAL638870-1G
SIAL638870-5G
2-Chloro-4-cyanobenzenesulphonyl chloride, Sigma-Aldrich®
2-Chloro-4-cyanobenzenesulphonyl chloride
Vzorec:
C₇H₃Cl₂NO₂S MW: 236,08 g/mol |
MDL Number:
MFCD00177089 CAS číslo: 254749-11-6 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...