Ethyl-2-chloroacetoacetate 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl 2-chloroacetoacetate
A19810.22
A19810.36
A19810.22EA
1380
CZK
A19810.22
A19810.36
Ethyl-2-chloroacetoacetate 96%
Ethyl-2-chloroacetoacetate
Purity: 96%
CAS No.: 609-15-4
Molecular Formula: C6H9ClO3
Formula Weight: 164.59
Flash Point: 82°(180°F)
Refractive Index: 1.4425
Melting Point:
Vzorec:
C₆H₉ClO₃ MW: 164,59 g/mol Bod varu: 200 °C Bod tání: < –80 °C Hustota: 1,182 Bod vzplanutí: 54 °C (180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009141 CAS číslo: 609-15-4 EINECS: 210-180-4 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...