2-Chloro-1-methylpyridinium iodide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
198005-100G
198005-10G
198005-25G
SIAL198005-100GEA
5420
CZK
SIAL198005-100G
SIAL198005-10G
SIAL198005-25G
2-Chloro-1-methylpyridinium iodide, Sigma-Aldrich®
2-Chloro-1-methylpyridinium iodide
Vzorec:
C₆H₇ClIN MW: 255,49 g/mol Bod tání: 207 °C |
MDL Number:
MFCD00011984 CAS číslo: 14338-32-0 EINECS: 238-288-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...