2-Chloro-1-methylpyridinium iodide ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A12820.14
A12820.22
A12820.36
A12820.14EA
1590
CZK
A12820.14
A12820.22
A12820.36
2-Chloro-1-methylpyridinium iodide ≥97%
2-Chloro-1-methylpyridinium iodide
Purity: 97%
CAS No.: 14338-32-0
Molecular Formula: C6H7ClIN
Formula Weight: 255.49
Melting Point: ca 200° dec.
Beilstein Reference: 3572320
Merck Reference: 14,6301
MDL No.: MFCD00011984
Vzorec:
C₆H₇ClIN MW: 255,49 g/mol Bod tání: ∼ 200 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011984 CAS číslo: 14338-32-0 EINECS: 238-288-7 Merck Index: 14,6301 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...