Tisk…

2-Butylfuran ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A13376.09 A13376.18 A13376.30
A13376.09EA 1950 CZK
A13376.09 A13376.18 A13376.30
2-Butylfuran ≥98%
2-Butylfuran

Purity: 98% CAS No.: 4466-24-4 Molecular Formula: C8H12O Formula Weight: 124.18 Storage Temperature: 2°C to 8°C Flash Point: 32°(89°F) Odour: Pleasant Boiling Point: 153-154° Beilstein Reference: 107045 UN No.: 1993 MDL No.: MFCD00047071

Vzorec: C₈H₁₂O
Bod varu: 153…154 °C
Hustota: 0,886
Bod vzplanutí: 32 °C (89 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00047071
CAS číslo: 4466-24-4
EINECS: 224-732-7
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR