Tisk…

2-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H26109.MD H26109.03
H26109.MDEA 889 CZK
H26109.MD H26109.03
2-Bromo-5-(trifluoromethyl)benzaldehyde ≥97%
2-Bromo-5-(trifluoromethyl)benzaldehyde

Purity: 98% CAS No.: 102684-91-3 Molecular Formula: C8H4BrF3O Formula Weight: 253.02 Refractive Index: 1.511

Vzorec: C₈H₄BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD04973760
CAS číslo: 102684-91-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR