Tisk…

2-Bromo-4,6-difluorobenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

555924-1G 555924-5G
SIAL555924-1GEA 1530 CZK
SIAL555924-1G SIAL555924-5G
2-Bromo-4,6-difluorobenzenesulphonyl chloride, Sigma-Aldrich®
2-Bromo-4,6-difluorobenzenesulphonyl chloride
Vzorec: C₆H₂BrClF₂O₂S
MW: 291,5 g/mol
Bod varu: 265…267 °C (1013 hPa)
Hustota: 1,93 g/cm³ (20 °C)
MDL Number: MFCD03094561
CAS číslo: 351003-42-4
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR