2-Bromo-4,5-dichloroimidazole ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Bromo-4,5-dichloro-1H-imidazole
A14896.03
A14896.06
A14896.03EA
1740
CZK
A14896.03
A14896.06
2-Bromo-4,5-dichloroimidazole ≥98%
2-Bromo-4,5-dichloroimidazole
Purity: 98%
CAS No.: 16076-27-0
Molecular Formula: C3HBrCl2N2
Formula Weight: 215.87
Odour: Odourless
Melting Point: ca 215° dec.
Beilstein Reference: 606732
MDL No.: MFCD00067894
Vzorec:
C₃HBrCl₂N₂ MW: 215,86 g/mol Bod tání: ∼ 215 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00067894 CAS číslo: 16076-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...