Tisk…

2-Aminobenzenesulphonamide 98%

Dodavatel: Thermo Scientific

B21912.06 B21912.14 B21912.22
B21912.06EA 1810 CZK
B21912.06 B21912.14 B21912.22
2-Aminobenzenesulphonamide 98%
2-Aminobenzenesulphonamide

Purity: 98% CAS No.: 3306-62-5 Molecular Formula: C6H8N2O2S Formula Weight: 172.21 Odour: Odourless Melting Point: 153-157° MDL No.: MFCD00007932

Vzorec: C₆H₈N₂O₂S
MW: 172,21 g/mol
Bod tání: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00007932
CAS číslo: 3306-62-5
EINECS: 221-988-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR