Tisk…

2-Aminobenzimidazole ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole

L02066.06 L02066.14 L02066.22
L02066.06EA 985 CZK
L02066.06 L02066.14 L02066.22
2-Aminobenzimidazole ≥97%
2-Aminobenzimidazole

Purity: 97+% CAS No.: 934-32-7 Molecular Formula: C7H7N3 Formula Weight: 133.15 Melting Point: 228-232° Beilstein Reference: 116525

Vzorec: C₇H₇N₃
MW: 133,15 g/mol
Bod varu: 381 °C (1013 hPa)
Bod tání: 228…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00005596
CAS číslo: 934-32-7
EINECS: 213-280-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR