2-Aminobenzimidazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1H-Benzo[d]imidazol-2-amine, Benzimidazol-2-ylamine, 2-Amino-1H-benzimidazole
M-Clarity™ quality level = MQ100
171778-25G
171778-5G
SIAL171778-25GEA
3700
CZK
SIAL171778-25G
SIAL171778-5G
2-Aminobenzimidazole, Sigma-Aldrich®
2-Aminobenzimidazole
Vzorec:
C₇H₇N₃ MW: 133,15 g/mol Bod varu: 381 °C (1013 hPa) Bod tání: 226…231 °C |
MDL Number:
MFCD00005596 CAS číslo: 934-32-7 EINECS: 213-280-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...