Tisk…

2,5-Dibromo-3-picoline, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,5-Dibromo-3-methylpyridine

M-Clarity™ quality level = MQ100

637661-1G 637661-5G
SIAL637661-1GEA 1130 CZK
SIAL637661-1G SIAL637661-5G
2,5-Dibromo-3-picoline, Sigma-Aldrich®
2,5-Dibromo-3-picoline
Vzorec: C₆H₅Br₂N
MW: 250,92 g/mol
MDL Number: MFCD02093085
CAS číslo: 3430-18-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR