Tisk…

2,4-Difluoro-5-nitrobenzoic acid ≥97%

Dodavatel: Thermo Scientific
Varování

H32617.06 H32617.14
H32617.06EA 3140 CZK
H32617.06 H32617.14
2,4-Difluoro-5-nitrobenzoic acid ≥97%
2,4-Difluoro-5-nitrobenzoic acid

Purity: 97% CAS No.: 153775-33-8 Molecular Formula: C7H3F2NO4 Formula Weight: 203.10 Melting Point: 125-133° MDL No.: MFCD04115713

Vzorec: C₇H₃F₂NO₄
Bod tání: 125…133 °C
Storage Temperature: Ambient
MDL Number: MFCD04115713
CAS číslo: 153775-33-8
UN: 2811
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR