Tisk…

2,4-Dichloro-5-methylbenzenesulphonyl chloride ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A19718.09 A19718.18 A19718.88
A19718.09EA 5530 CZK
A19718.09 A19718.18 A19718.88
2,4-Dichloro-5-methylbenzenesulphonyl chloride ≥97%
2,4-Dichloro-5-methylbenzenesulphonyl chloride

Purity: 97% CAS No.: 28286-86-4 Molecular Formula: C7H5Cl3O2S Formula Weight: 259.54 Melting Point: 70-73° UN No.: 3261 MDL No.: MFCD00044016

Vzorec: C₇H₅Cl₃O₂S
Bod tání: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00044016
CAS číslo: 28286-86-4
EINECS: 248-938-1
UN: 3261
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR