Tisk…

2,4,6-Trifluorobenzenesulphonyl chloride ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2,4,6-Trifluorobenzene-1-sulphonyl chloride

H32746.MD H32746.03
H32746.MDEA 1960 CZK
H32746.MD H32746.03
2,4,6-Trifluorobenzenesulphonyl chloride ≥97%
2,4,6-Trifluorobenzenesulphonyl chloride

Purity: 97% CAS No.: 220239-64-5 Molecular Formula: C6H2ClF3O2S Formula Weight: 230.59 UN No.: 3265 MDL No.: MFCD01091015

Vzorec: C₆H₂ClF₃O₂S
MW: 230,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01091015
CAS číslo: 220239-64-5
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR