Mucic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Galactaric acid, rel-(2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid, (2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid
M-Clarity™ quality level = MQ200
M89617-100G
M89617-500G
SIALM89617-100GEA
1790
CZK
SIALM89617-100G
SIALM89617-500G
Mucic acid, Sigma-Aldrich®
Mucic acid
Vzorec:
C₆H₁₀O₈ MW: 210,14 g/mol Bod tání: 220…225 °C Hustota: 1,79 g/cm³ (20 °C) |
MDL Number:
MFCD00004239 CAS číslo: 526-99-8 EINECS: 208-404-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...