Tisk…

Mucic acid ≥98%

Dodavatel: Thermo Scientific

Synonyms: Galactaric acid, rel-(2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid, (2R,3S,4R,5S)-2,3,4,5-Tetrahydroxyhexanedioic acid

B24170.22 B24170.36
B24170.22EA 1620 CZK
B24170.22 B24170.36
Mucic acid ≥98%
Mucic acid

Purity: 98% CAS No.: 526-99-8 Molecular Formula: C6H10O8 Formula Weight: 210.14 Melting Point: ca 210° dec. Beilstein Reference: 1728117 Merck Reference: 14,4331 MDL No.: MFCD00004239

Vzorec: C₆H₁₀O₈
MW: 210,14 g/mol
Bod tání: ∼ 210 °C (rozkl.)
Hustota: 1,79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004239
CAS číslo: 526-99-8
EINECS: 208-404-0
Merck Index: 14,4331

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR