2-(2-Hydroxyethyl)pyridine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Pyridineethanol, 2-(2-Pyridyl)ethanol
M-Clarity™ quality level = MQ100
128643-100G
128643-500G
128643-5G
SIAL128643-100GEA
3390
CZK
SIAL128643-100G
SIAL128643-500G
SIAL128643-5G
2-(2-Hydroxyethyl)pyridine, Sigma-Aldrich®
2-(2-Hydroxyethyl)pyridine
Vzorec:
C₇H₉NO MW: 123,15 g/mol Bod varu: 241 °C (1013 hPa) Bod tání: –7,8 °C Hustota: 1,1111 g/cm³ (4 °C) |
MDL Number:
MFCD00006364 CAS číslo: 103-74-2 EINECS: 203-140-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...