2-Nitro-4-(trifluoromethyl)phenol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L10604.03
L10604.06
L10604.03EA
1910
CZK
L10604.03
L10604.06
2-Nitro-4-(trifluoromethyl)phenol ≥98%
2-Nitro-4-(trifluoromethyl)phenol
Purity: 98%
CAS No.: 400-99-7
Molecular Formula: C7H4F3NO3
Formula Weight: 207.11
Flash Point: 95°(203°F)
Refractive Index: 1.5040
Boiling Point: 92-94°/12mm
Beilstein Reference: 2215510
MDL No.: MFCD00009791
Vzorec:
C₇H₄F₃NO₃ Bod varu: 92…94 °C (12 mmHg) Hustota: 1,473 Bod vzplanutí: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009791 CAS číslo: 400-99-7 EINECS: 206-927-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...