4-Nitro-3-(trifluoromethyl)phenol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L08228.03
L08228.06
L08228.03EA
1030
CZK
L08228.03
L08228.06
4-Nitro-3-(trifluoromethyl)phenol ≥97%
4-Nitro-3-(trifluoromethyl)phenol
Purity: 97%
CAS No.: 88-30-2
Molecular Formula: C7H4F3NO3
Formula Weight: 207.11
Melting Point: 74-77°
Boiling Point: 135-138°/0.01mm
Merck Reference: 14,9253
UN No.: 2811
MDL No.: MFCD00014714
Vzorec:
C₇H₄F₃NO₃ Bod varu: 135…138 °C (0,01 mmHg) Bod tání: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014714 CAS číslo: 88-30-2 EINECS: 201-818-2 UN: 2811 ADR: 6.1,III Merck Index: 14,9253 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...