Tisk…

1-Ethylpyridinium bromide ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Ethylpyridin-1-ium bromide

A17911.14 A17911.22
A17911.14EA 1570 CZK
A17911.14 A17911.22
1-Ethylpyridinium bromide ≥99%
1-Ethylpyridinium bromide

Purity: 99% CAS No.: 1906-79-2 Molecular Formula: C7H10BrN Formula Weight: 188.07 Melting Point: 117-121° MDL No.: MFCD00041999

Vzorec: C₇H₁₀BrN
MW: 188,07 g/mol
Bod tání: 117…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00041999
CAS číslo: 1906-79-2
EINECS: 217-607-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR