3-Ethyl-2-iodotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
1-Ethyl-2-iodo-3-methylbenzene, 2-Ethyl-6-methyliodobenzene
A10839.06
A10839.14
A10839.06EA
1710
CZK
A10839.06
A10839.14
3-Ethyl-2-iodotoluene ≥98%
3-Ethyl-2-iodotoluene
Purity: 98%
CAS No.: 175277-95-9
Molecular Formula: C9H11I
Formula Weight: 246.08
Boiling Point: 118-120°/15mm
MDL No.: MFCD00052855
Vzorec:
C₉H₁₁I MW: 246.09 Bod varu: 118…120 °C (15 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00052855 CAS číslo: 175277-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...