4-Hydroxy-3,5-dijodbenzonitril ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Ioxynil
A13929.06
A13929.14
A13929.06EA
2520
CZK
A13929.06
A13929.14
4-Hydroxy-3,5-dijodbenzonitril ≥97%
4-Hydroxy-3,5-dijodbenzonitril
Purity: 97%
CAS No.: 1689-83-4
Molecular Formula: C7H3I2NO
Formula Weight: 370.92
Odour: Odourless
Melting Point: 200-207°
Beilstein Reference: 2364041
UN No.: 3439
MDL No.: MFCD00016422
Vzorec:
C₇H₃I₂NO MW: 370,92 g/mol Bod varu: 378 °C (1013 hPa) Bod tání: 200…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016422 CAS číslo: 1689-83-4 EINECS: 216-881-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...