Tisk…

2-Chlorfenol-D4, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

449423-100MG 449423-250MG
SIAL449423-100MGEA 6420 CZK
SIAL449423-100MG SIAL449423-250MG
2-Chlorfenol-D4, Sigma-Aldrich®
2-Chlorfenol-D4
Vzorec: C₆HD₄ClO
Storage Temperature: Ambient
CAS číslo: 93951-73-6
UN: 2021
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR