Tisk…

(2-Chlorophenyl)acetylene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Chloro-2-ethynylbenzene

H53441.03 H53441.06
H53441.03EA 2880 CZK
H53441.03 H53441.06
(2-Chlorophenyl)acetylene ≥98%
(2-Chlorophenyl)acetylene

Purity: 98% CAS No.: 873-31-4 Molecular Formula: C8H5Cl Formula Weight: 136.58 Flash Point: 70°(158°F) Refractive Index: 1.5710 Boiling Point: 82°/12mm MDL No.: MFCD00269951

Vzorec: C₈H₅Cl
MW: 136,58 g/mol
Bod varu: 82 °C (12 mmHg)
Hustota: 1,125
Bod vzplanutí: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00269951
CAS číslo: 873-31-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR