4'-Butylacetophenone ≥97%
Dodavatel: Thermo Scientific
Synonyms:
4'-n-Butylacetophenone, 4-Butylacetophenone, p-Butylacetophenone
A15296.06
A15296.14
A15296.06EA
1800
CZK
A15296.06
A15296.14
4'-Butylacetophenone ≥97%
4'-Butylacetophenone
Purity: 97%
CAS No.: 37920-25-5
Molecular Formula: C12H16O
Formula Weight: 176.26
Flash Point:
Vzorec:
C₁₂H₁₆O MW: 176,26 g/mol Bod varu: 101…102 °C (1,5 mmHg) Hustota: 0,957 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017500 CAS číslo: 37920-25-5 EINECS: 253-715-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...