Tisk…

4-Bromo-2-fluorobenzonitrile ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

A13086.06 A13086.14
A13086.06EA 1150 CZK
A13086.06 A13086.14
4-Bromo-2-fluorobenzonitrile ≥99%
4-Bromo-2-fluorobenzonitrile

Purity: 99% CAS No.: 105942-08-3 Molecular Formula: C7H3BrFN Formula Weight: 200.01 Melting Point: 69-72° Boiling Point: 238-239° UN No.: 3439 MDL No.: MFCD00143264

Vzorec: C₇H₃BrFN
Bod varu: 238…239 °C
Bod tání: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00143264
CAS číslo: 105942-08-3
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR