3-Bromo-4-hydroxybenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
A19400.06
A19400.14
A19400.06EA
1580
CZK
A19400.06
A19400.14
3-Bromo-4-hydroxybenzonitrile ≥98%
3-Bromo-4-hydroxybenzonitrile
Purity: 98%
CAS No.: 2315-86-8
Molecular Formula: C7H4BrNO
Formula Weight: 198.02
Melting Point: 156-161°
Beilstein Reference: 2207020
UN No.: 3439
MDL No.: MFCD00143096
Vzorec:
C₇H₄BrNO MW: 198,02 g/mol Bod varu: 406 °C (1013 hPa) Bod tání: 156…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143096 CAS číslo: 2315-86-8 EINECS: 219-022-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...