5-Bromo-2-fluorobenzonitrile, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
465259-1G
465259-5G
SIAL465259-1GEA
707
CZK
SIAL465259-1G
SIAL465259-5G
5-Bromo-2-fluorobenzonitrile, Sigma-Aldrich®
5-Bromo-2-fluorobenzonitrile
Vzorec:
C₇H₃BrFN MW: 200,01 g/mol Bod varu: 255 °C (1013 hPa) Bod tání: 75…80 °C |
MDL Number:
MFCD00143424 CAS číslo: 179897-89-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...