Tisk…

5-Bromo-2-fluorobenzonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A16113.14 A16113.06
A16113.14EA 3080 CZK
A16113.14 A16113.06
5-Bromo-2-fluorobenzonitrile ≥98%
5-Bromo-2-fluorobenzonitrile

Purity: 98% CAS No.: 179897-89-3 Molecular Formula: C7H3BrFN Formula Weight: 200.01 Melting Point: 77-81° Beilstein Reference: 7701063 UN No.: 3439 MDL No.: MFCD00143424

Vzorec: C₇H₃BrFN
Bod varu: 255 °C (1013 hPa)
Bod tání: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00143424
CAS číslo: 179897-89-3
UN: 3439
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR