Tisk…

4-Bromo-3-fluorophenol ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

H32488.06 H32488.14
H32488.06EA 1860 CZK
H32488.06 H32488.14
4-Bromo-3-fluorophenol ≥98%
4-Bromo-3-fluorophenol

Purity: 98% CAS No.: 121219-03-2 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Melting Point: 71-73° MDL No.: MFCD00051907

Vzorec: C₆H₄BrFO
Bod tání: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00051907
CAS číslo: 121219-03-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR