Tisk…

2-Amino-5-nitrobenzophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2-Amino-5-nitrophenyl)(phenyl)methanone

M-Clarity™ quality level = MQ100

211737-100G 211737-25G
SIAL211737-100GEA 6040 CZK
SIAL211737-100G SIAL211737-25G
2-Amino-5-nitrobenzophenone, Sigma-Aldrich®
2-Amino-5-nitrobenzophenone
Vzorec: C₁₃H₁₀N₂O₃
MW: 242,23 g/mol
Bod tání: 163,5 °C
Hustota: 1,474 g/cm³ (20 °C)
MDL Number: MFCD00007364
CAS číslo: 1775-95-7
EINECS: 217-207-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR