Hydrocortisone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
H4001-1G
H4001-100G
H4001-10G
H4001-25G
H4001-5G
SIALH4001-1GEA
2030
CZK
SIALH4001-1G
SIALH4001-100G
SIALH4001-10G
SIALH4001-25G
SIALH4001-5G
Hydrocortisone, Sigma-Aldrich®
Hydrocortisone
Vzorec:
C₂₁H₃₀O₅ Bod varu: 567 °C (1013 hPa) Bod tání: 202…204 °C Hustota: 1,4 g/cm³ (20 °C) |
MDL Number:
MFCD00011654 CAS číslo: 50-23-7 EINECS: 200-020-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...