Hydrocortisone, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
H3160-10G
H3160-25G
H3160-5G
SIALH3160-10GEA
18330
CZK
SIALH3160-10G
SIALH3160-25G
SIALH3160-5G
Hydrocortisone, Sigma-Aldrich®
Hydrocortisone
Vzorec:
C₂₁H₃₀O₅ MW: 362.47 Bod varu: 567 Bod tání: 202…204 °C Hustota: 1.4 g/cm³ (20 °C) |
MDL Number:
MFCD00011654 CAS číslo: 50-23-7 EINECS: 200-020-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...