1,4-Diaminobutane dihydrochloride 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1,4-Butanediamine dihydrochloride, Tetramethylenediammonium dichloride, Tetramethylendiamine dihydrochloride, Putrescine dihydrochloride
A18312.22
A18312.14
A18312.22EA
6560
CZK
A18312.22
A18312.14
1,4-Diaminobutane dihydrochloride 99%
1,4-Diaminobutane dihydrochloride
Purity: 99%
CAS No.: 333-93-7
Molecular Formula: HCl.H2N(CH2)4NH2.HCl
Formula Weight: 161.08
Melting Point: ca 280° dec.
Beilstein Reference: 3906680
Merck Reference: 14,7947
MDL No.: MFCD00012526
Vzorec:
C₄H₁₄Cl₂N₂ MW: 161,07 g/mol Bod tání: ∼ 280 °C (rozkl.) Hustota: 1,31 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012526 CAS číslo: 333-93-7 EINECS: 206-375-9 Merck Index: 14,7947 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...