Tisk…

3',5'-Dimethoxy-4'-hydroxyacetophenone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3′,5′-Dimethoxy-4′-hydroxyacetophenone, 4'-Hydroxy-3',5'-dimethoxyacetophenone, 1-(3,5-Dimethoxy-4-hydroxyphenyl)ethan-1-one, Acetosyringone, 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanone

M-Clarity™ quality level = MQ100

D134406-5G D134406-1G D134406-25G
SIALD134406-5GEA 2950 CZK
SIALD134406-5G SIALD134406-1G SIALD134406-25G
3',5'-Dimethoxy-4'-hydroxyacetophenone, Sigma-Aldrich®
3',5'-Dimethoxy-4'-hydroxyacetophenone
Vzorec: C₁₀H₁₂O₄
MW: 196,2 g/mol
Bod varu: 335 °C (1013 hPa)
Bod tání: 119…120 °C
MDL Number: MFCD00008748
CAS číslo: 2478-38-8
EINECS: 219-610-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR